Assay Details
| Gene Symbol: Mrpl57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500422.2 | XP_003500470.1 | 2 | 291 | 98 | |
| XM_007641162.1 | XP_007639352.1 | 2 | 225 | 98 | ||
| XM_007641163.1 | XP_007639353.1 | 1 | 216 | 98 | ||
More Information
Set Membership: |
|

Mrpl57
Genomic Map