Assay Details
Gene Symbol: B3galt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510834.2 | XP_003510882.2 | 1 | 173 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
