Assay Details
Gene Symbol: Mal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497801.1 | XP_003497849.1 | 1 | 22 | 66 | |
XM_003497802.1 | XP_003497850.1 | 1 | 22 | 66 | ||
XM_007617928.1 | XP_007616118.1 | 1 | 22 | 66 | ||
XM_007617944.1 | XP_007616134.1 | 1 | 22 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
