Assay Details
| Gene Symbol: Hoxc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507216.1 | XP_003507264.1 | 2 | 221 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxc12| Gene Symbol: Hoxc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507216.1 | XP_003507264.1 | 2 | 221 | 108 | |
Set Membership: |
|