Assay Details
| Gene Symbol: ZC3H15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675879.2 | XP_005675936.1 | 3 - 4 | 516 | 79 | |
| GenBank mRNA | JO611998.2 | - | 7 - 8 | 1609 | 79 | |
More Information
Set Membership: |
|

ZC3H15
Genomic Map