Assay Details
| Gene Symbol: PEA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046375.1 | XP_017901864.1 | 3 - 4 | 494 | 73 | |
| GenBank mRNA | JO355652.2 | - | 3 - 4 | 437 | 73 | |
| JO557227.2 | - | 3 - 4 | 473 | 73 | ||
| JO633971.2 | - | 3 - 4 | 455 | 73 | ||
More Information
Set Membership: |
|

PEA15
Genomic Map