Assay Details
Gene Symbol: CAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679159.2 | XP_005679216.1 | 2 - 3 | 491 | 67 | |
XM_005679160.3 | XP_005679217.1 | 2 - 3 | 346 | 67 | ||
XM_005679161.2 | XP_005679218.1 | 2 - 3 | 279 | 67 | ||
GenBank mRNA | DQ912799.1 | - | 1 - 2 | 173 | 67 | |
JO642931.2 | - | 1 - 2 | 594 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
