Assay Details
| Gene Symbol: LRP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054565.1 | XP_017910054.1 | 4 - 5 | 1305 | 71 | |
| XM_018054566.1 | XP_017910055.1 | 3 - 4 | 412 | 71 | ||
| GenBank mRNA | JO425256.1 | - | 4 - 5 | 1057 | 71 | |
More Information
Set Membership: |
|

LRP10
Genomic Map