Assay Details
Gene Symbol: LOC102175297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685071.3 | XP_005685128.1 | 3 - 4 | 501 | 80 | |
GenBank mRNA | JO419757.2 | - | 5 - 6 | 1497 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
