Assay Details
Gene Symbol: RNASEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693473.3 | XP_005693530.1 | 2 - 3 | 284 | 69 | |
GenBank mRNA | JO568344.2 | - | 1 - 2 | 399 | 69 | |
JO612686.2 | - | 1 - 2 | 380 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
