Assay Details
Gene Symbol: EMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695616.3 | XP_005695673.1 | 1 - 2 | 393 | 60 | |
GenBank mRNA | JO427054.1 | - | 1 - 2 | 406 | 60 | |
KA337968.1 | - | 7 - 8 | 2417 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
