Assay Details
| Gene Symbol: CCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685708.2 | XP_005685765.1 | 5 - 6 | 769 | 63 | |
| GenBank mRNA | JO592745.2 | - | 4 - 5 | 749 | 63 | |
| KA350371.1 | - | 5 - 6 | 716 | 63 | ||
More Information
Set Membership: |
|

CCNB2
Genomic Map