Assay Details
| Gene Symbol: SLC6A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044781.1 | XP_017900270.1 | 6 - 7 | 1674 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC6A8| Gene Symbol: SLC6A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044781.1 | XP_017900270.1 | 6 - 7 | 1674 | 63 | |
Set Membership: |
|