Assay Details
| Gene Symbol: ARPP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013967132.2 | XP_013822586.2 | 2 - 3 | 230 | 81 | |
| XM_018054219.1 | XP_017909708.1 | 3 - 4 | 261 | 81 | ||
| GenBank mRNA | JO366734.2 | - | 1 - 2 | 475 | 81 | |
| KA329051.1 | - | 3 - 4 | 371 | 81 | ||
More Information
Set Membership: |
|

ARPP19
Genomic Map