Assay Details
| Gene Symbol: GRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042871.1 | XP_017898360.1 | 13 - 14 | 1406 | 55 | |
| GenBank mRNA | JO579500.1 | - | 13 - 14 | 1244 | 55 | |
| JO643899.2 | - | 8 - 9 | 2037 | 55 | ||
More Information
Set Membership: |
|

GRK2
Genomic Map