Assay Details
| Gene Symbol: MYL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680364.3 | XP_005680421.1 | 4 - 5 | 449 | 67 | |
| XM_005680365.2 | XP_005680422.1 | 5 - 6 | 561 | 67 | ||
| GenBank mRNA | JO632191.2 | - | 2 - 3 | 247 | 67 | |
More Information
Set Membership: |
|

MYL6
Genomic Map