Assay Details
| Gene Symbol: NDUFS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694716.2 | XP_005694773.1 | 1 - 2 | 131 | 60 | |
| GenBank mRNA | JO516244.2 | - | 1 - 2 | 143 | 60 | |
More Information
Set Membership: |
|

NDUFS4
Genomic Map