Assay Details
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285608.1 | NP_001272537.1 | 4 - 5 | 663 | 57 | |
XM_013963900.1 | XP_013819354.1 | 4 - 5 | 728 | 57 | ||
GenBank mRNA | EF035041.1 | - | 4 - 5 | 663 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
