Assay Details
| Gene Symbol: MAPK1IP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685883.3 | XP_005685940.2 | 3 - 4 | 885 | 89 | |
| GenBank mRNA | JO576702.1 | - | 2 - 3 | 923 | 89 | |
More Information
Set Membership: |
|

MAPK1IP1L
Genomic Map