Assay Details
Gene Symbol: C17H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691577.3 | XP_005691634.2 | 3 - 4 | 304 | 75 | |
XM_005691578.3 | XP_005691635.2 | 3 - 4 | 300 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
