Assay Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044721.1 | XP_017900210.1 | 12 - 13 | 2511 | 58 | |
| XM_018044722.1 | XP_017900211.1 | 11 - 12 | 2244 | 58 | ||
More Information
Set Membership: |
|

CLCN4
Genomic Map