Assay Details
| Gene Symbol: TUBB4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050940.1 | XP_017906429.1 | 4 - 5 | 522 | 81 | |
| GenBank mRNA | JO420802.2 | - | 1 - 2 | 1651 | 81 | |
More Information
Set Membership: |
|

TUBB4A
Genomic Map