Assay Details
Gene Symbol: SH3BGRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056334.1 | XP_017911823.1 | 2 - 3 | 516 | 66 | |
GenBank mRNA | JO349203.2 | - | 2 - 3 | 342 | 66 | |
JO632773.2 | - | 2 - 3 | 377 | 66 | ||
KA308073.1 | - | 1 - 2 | 474 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
