Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962463.2 | XP_013817917.1 | 3 - 4 | 268 | 73 | |
XM_018046014.1 | XP_017901503.1 | 3 - 4 | 265 | 73 | ||
GenBank mRNA | JO581709.1 | - | 3 - 4 | 1668 | 73 | |
KA334559.1 | - | 3 - 4 | 454 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
