Assay Details
| Gene Symbol: SORD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685826.3 | XP_005685883.2 | 4 - 5 | 563 | 73 | |
| GenBank mRNA | JO421558.2 | - | 4 - 5 | 705 | 73 | |
| JO427553.1 | - | 4 - 5 | 625 | 73 | ||
More Information
Set Membership: |
|

SORD
Genomic Map