Assay Details
Gene Symbol: AMH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050765.1 | XP_017906254.1 | 2 - 3 | 1177 | 87 | |
XM_018050766.1 | XP_017906255.1 | 2 - 3 | 1177 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
