Assay Details
| Gene Symbol: PRDX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678534.3 | XP_005678591.1 | 5 - 6 | 655 | 67 | |
| GenBank mRNA | JO535987.2 | - | 5 - 6 | 623 | 67 | |
More Information
Set Membership: |
|

PRDX1
Genomic Map