Assay Details
| Gene Symbol: LOC102189116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682625.3 | XP_005682682.1 | 11 - 12 | 1533 | 69 | |
| XM_013965331.2 | XP_013820785.1 | 9 - 10 | 1265 | 69 | ||
| XM_013965333.2 | XP_013820787.1 | 9 - 10 | 1265 | 69 | ||
| XM_018050726.1 | XP_017906215.1 | 11 - 12 | 1532 | 69 | ||
| XM_018050727.1 | XP_017906216.1 | 11 - 12 | 1602 | 69 | ||
| XM_018050728.1 | XP_017906217.1 | 12 - 13 | 1637 | 69 | ||
| XM_018050729.1 | XP_017906218.1 | 11 - 12 | 1523 | 69 | ||
| XM_018050730.1 | XP_017906219.1 | 11 - 12 | 1532 | 69 | ||
| GenBank mRNA | JO427600.1 | - | 14 - 15 | 1939 | 69 | |
More Information
Set Membership: |
|

LOC102189116
Genomic Map