Assay Details
Gene Symbol: CIDEB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685207.3 | XP_005685264.1 | 2 - 3 | 950 | 77 | |
XM_018054643.1 | XP_017910132.1 | 2 - 3 | 257 | 77 | ||
XM_018054644.1 | XP_017910133.1 | 2 - 3 | 247 | 77 | ||
GenBank mRNA | JO437600.2 | - | 3 - 4 | 930 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
