Assay Details
| Gene Symbol: DDX19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061686.1 | XP_017917175.1 | 2 - 3 | 239 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX19A| Gene Symbol: DDX19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061686.1 | XP_017917175.1 | 2 - 3 | 239 | 99 | |
Set Membership: |
|