Assay Details
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005688259.3 | XP_005688316.1 | 8 - 9 | 754 | 78 | |
XM_018057495.1 | XP_017912984.1 | 7 - 8 | 692 | 78 | ||
XM_018057496.1 | XP_017912985.1 | 6 - 7 | 683 | 78 | ||
XM_018057497.1 | XP_017912986.1 | 5 - 6 | 621 | 78 | ||
GenBank mRNA | JO501202.2 | - | 6 - 7 | 782 | 78 | |
KA347403.1 | - | 1 - 2 | 1746 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
