Assay Details
Gene Symbol: LAMTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677361.3 | XP_005677418.1 | 3 - 4 | 474 | 60 | |
GenBank mRNA | JT657754.1 | - | 1 - 2 | 142 | 60 | |
KA349045.1 | - | 3 - 4 | 403 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
