Assay Details
| Gene Symbol: RIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696773.3 | XP_005696830.1 | 10 - 11 | 961 | 58 | |
| XM_005696774.2 | XP_005696831.1 | 10 - 11 | 1081 | 58 | ||
| XM_018039210.1 | XP_017894699.1 | 10 - 11 | 960 | 58 | ||
More Information
Set Membership: |
|

RIPOR2
Genomic Map