Assay Details
Gene Symbol: FKBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699849.2 | XP_005699906.1 | 3 - 4 | 470 | 74 | |
XM_005699850.3 | XP_005699907.1 | 3 - 4 | 412 | 74 | ||
XM_005699851.3 | XP_005699908.1 | 3 - 4 | 409 | 74 | ||
XM_018042788.1 | XP_017898277.1 | 3 - 4 | 760 | 74 | ||
GenBank mRNA | JI861823.1 | - | 2 - 3 | 74 | ||
JO452042.2 | - | 3 - 4 | 224 | 74 | ||
KA322925.1 | - | 3 - 4 | 440 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
