Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697495.2 | XP_005697552.1 | 3 - 4 | 374 | 69 | |
| GenBank mRNA | JO413800.2 | - | 2 - 3 | 272 | 69 | |
| JO581637.1 | - | 1 - 2 | 579 | 69 | ||
More Information
Set Membership: |
|

CARHSP1
Genomic Map