Assay Details
Gene Symbol: B4GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041313.1 | XP_017896802.1 | 5 - 6 | 1053 | 61 | |
XM_018041382.1 | XP_017896871.1 | 6 - 7 | 1111 | 61 | ||
XM_018041442.1 | XP_017896931.1 | 6 - 7 | 1078 | 61 | ||
GenBank mRNA | JO335650.2 | - | 1 - 2 | 157 | 61 | |
JO361670.2 | - | 2 - 3 | 388 | 61 | ||
JO418395.2 | - | 2 - 3 | 258 | 61 | ||
JO430717.2 | - | 2 - 3 | 985 | 61 | ||
JO439131.2 | - | 2 - 3 | 985 | 61 | ||
JO523096.2 | - | 1 - 2 | 173 | 61 | ||
JO549279.2 | - | 2 - 3 | 330 | 61 | ||
JO566339.2 | - | 2 - 3 | 389 | 61 | ||
JO575884.2 | - | 2 - 3 | 389 | 61 | ||
JO594624.2 | - | 2 - 3 | 1009 | 61 | ||
JO594625.2 | - | 2 - 3 | 1009 | 61 | ||
JO615581.2 | - | 2 - 3 | 290 | 61 | ||
KA337326.1 | - | 5 - 6 | 972 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
