Assay Details
| Gene Symbol: ARL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699871.3 | XP_005699928.1 | 2 - 3 | 558 | 62 | |
| GenBank mRNA | JO632991.2 | - | 2 - 3 | 298 | 62 | |
More Information
Set Membership: |
|

ARL2
Genomic Map