Assay Details
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693467.3 | XP_005693524.1 | 1 - 2 | 155 | 101 | |
GenBank mRNA | KA332080.1 | - | 2 - 3 | 988 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
