Assay Details
| Gene Symbol: DMRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692575.1 | XP_005692632.1 | 2 - 3 | 284 | 61 | |
| XM_005692576.3 | XP_005692633.1 | 1 - 2 | 318 | 61 | ||
More Information
Set Membership: |
|

DMRTC2
Genomic Map