Assay Details
Gene Symbol: ATP5PB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677897.3 | XP_005677954.1 | 5 - 6 | 603 | 66 | |
GenBank mRNA | JO437631.2 | - | 5 - 6 | 639 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
