Assay Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043302.1 | XP_017898791.1 | 4 - 5 | 688 | 81 | |
XM_018043303.1 | XP_017898792.1 | 4 - 5 | 683 | 81 | ||
XM_018043304.1 | XP_017898793.1 | 4 - 5 | 489 | 81 | ||
XM_018043305.1 | XP_017898794.1 | 3 - 4 | 391 | 81 | ||
XM_018043306.1 | XP_017898795.1 | 4 - 5 | 688 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
