Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061305.1 | XP_017916794.1 | 4 - 5 | 489 | 68 | |
XM_018061306.1 | XP_017916795.1 | 4 - 5 | 490 | 68 | ||
GenBank mRNA | JO401887.2 | - | 4 - 5 | 447 | 68 | |
JO577666.1 | - | 2 - 3 | 1054 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
