Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693590.2 | XP_005693647.2 | 3 - 4 | 307 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693590.2 | XP_005693647.2 | 3 - 4 | 307 | 64 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |