Assay Details
| Gene Symbol: DPY30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686393.2 | XP_005686450.1 | 2 - 3 | 178 | 85 | |
| XM_005686394.3 | XP_005686451.1 | 2 - 3 | 302 | 85 | ||
| GenBank mRNA | JO609267.2 | - | 2 - 3 | 154 | 85 | |
| KA304604.1 | - | 3 - 4 | 534 | 85 | ||
More Information
Set Membership: |
|

DPY30
Genomic Map