Assay Details
Gene Symbol: UBE2E3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013968525.2 | XP_013823979.2 | 3 - 4 | 429 | 99 | |
XM_018065310.1 | XP_017920799.1 | 3 - 4 | 664 | 99 | ||
XM_018065316.1 | XP_017920805.1 | 3 - 4 | 410 | 99 | ||
GenBank mRNA | JT725346.1 | - | 3 - 4 | 1034 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
