Assay Details
| Gene Symbol: GFOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692154.3 | XP_005692211.1 | 2 - 3 | 475 | 66 | |
| XM_005692155.3 | XP_005692212.1 | 2 - 3 | 451 | 66 | ||
| XM_018062170.1 | XP_017917659.1 | 2 - 3 | 1031 | 66 | ||
| GenBank mRNA | KA305988.1 | - | 1 - 2 | 1556 | 66 | |
More Information
Set Membership: |
|

GFOD2
Genomic Map