Assay Details
| Gene Symbol: CDKN2AIPNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683309.3 | XP_005683366.2 | 1 - 2 | 429 | 70 | |
| XM_018050385.1 | XP_017905874.1 | 1 - 2 | 429 | 70 | ||
| GenBank mRNA | JO612364.2 | - | 2 - 3 | 580 | 70 | |
| KA345189.1 | - | 1 - 2 | 310 | 70 | ||
More Information
Set Membership: |
|

CDKN2AIPNL
Genomic Map