Assay Details
Gene Symbol: VPS37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041919.1 | XP_017897408.1 | 4 - 5 | 733 | 62 | |
GenBank mRNA | JO410527.2 | - | 4 - 5 | 621 | 62 | |
JR131289.1 | - | 755 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
