Assay Details
| Gene Symbol: SLC10A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686021.2 | XP_005686078.2 | 2 - 3 | 890 | 64 | |
| GenBank mRNA | JO421963.2 | - | 2 - 3 | 769 | 64 | |
More Information
Set Membership: |
|

SLC10A1
Genomic Map