Assay Details
Gene Symbol: LRRC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044430.1 | XP_017899919.1 | 2 - 3 | 718 | 73 | |
GenBank mRNA | JO399063.2 | - | 1 - 2 | 87 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
